AMBH303C5CD3 Ambeed, Inc.

1-(4-Methoxyphenyl)ethylamine

  • MDL number MFCD00044523
  • Cas Number : 6298-96-0
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 99.3
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 240.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/6298-96-0.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/6298-96-0.html
pictograms GHS05
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart