AMBH303C5D0F Ambeed, Inc.

2,2-Dimethyltetrahydrothiazole

  • MDL number MFCD00014488
  • Cas Number : 19351-18-9
  • Molecular Weight : 117.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/19351-18-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/19351-18-9.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart