AMBH303C5D45 Ambeed, Inc.

Benzyl (1-benzyl-2,5-dioxopyrrolidin-3-yl)carbamate

  • MDL number MFCD12965073
  • Cas Number : 1219424-59-5
  • Molecular Weight : 338.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1219424-59-5.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1219424-59-5.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart