AMBH303C5D88 Ambeed, Inc.

Imidazo[1,2-a]pyrimidine-2-acetic acid hydrochloride

  • MDL number MFCD09763586
  • Cas Number : 1049730-74-6
  • Molecular Weight : 213.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1049730-74-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1049730-74-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart