AMBH303C5DBB Ambeed, Inc.

3-Isopropyl-5-nitropyridine

  • MDL number MFCD18803311
  • Cas Number : 131941-33-8
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/131941-33-8.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/131941-33-8.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart