AMBH303C5DEA Ambeed, Inc.

4-(4-Chlorophenyl)piperidine

  • MDL number MFCD03077010
  • Cas Number : 26905-02-2
  • Molecular Weight : 195.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/26905-02-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/26905-02-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart