AMBH303C5DFC Ambeed, Inc.

2-Chloro-5-fluoro-3-nitropyridin-4-amine

  • MDL number MFCD19441221
  • Cas Number : 405230-90-2
  • Molecular Weight : 191.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/405230-90-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/405230-90-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart