AMBH303C5E10 Ambeed, Inc.

1-Chloro-2-fluoro-3-methylbenzene

  • MDL number MFCD00042115
  • Cas Number : 85089-31-2
  • Molecular Weight : 144.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/85089-31-2.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/85089-31-2.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart