AMBH303C5E1F Ambeed, Inc.

Cefotaxime Sodium

  • MDL number MFCD00079073
  • Cas Number : 64485-93-4
  • Molecular Weight : 477.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-163
Synonym Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/64485-93-4.html

Safety Information

signalword Danger
pcodes P261-P280-P342+P311
hcodes 6.1
MSDS https://www.ambeed.com/products/64485-93-4.html
pictograms GHS08
hstatements H317-H334
Availability details for
Quantity
  • Availability
  • Add to Cart