AMBH303C5E2F Ambeed, Inc.

1,8-Dibromopyrene

  • MDL number MFCD00230868
  • Cas Number : 38303-35-4
  • Molecular Weight : 360.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-213�t N/A mmHg
Purity 97
Flash Point 277.3
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 469.6�t 760 mmHg
Product Link https://www.ambeed.com/products/38303-35-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/38303-35-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart