AMBH303C5E4C Ambeed, Inc.

(R)-2-(Pyrrolidin-2-yl)pyridine dihydrochloride

  • MDL number MFCD28041187
  • Molecular Weight : 221.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/P000457155.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/P000457155.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart