AMBH303C5E52 Ambeed, Inc.

3-(((2S,3R,4R,5R)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one

  • MDL number MFCD29050128
  • Cas Number : 21637-25-2
  • Molecular Weight : 464.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/21637-25-2.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/21637-25-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart