AMBH303C5E5C Ambeed, Inc.

6-Chloro-1H-indazole-4-carboxylic acid

  • MDL number MFCD07781757
  • Cas Number : 885522-12-3
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/885522-12-3.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/885522-12-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart