AMBH303C5E5E Ambeed, Inc.

(2S,3R,4S,5S,6R)-6-((Trityloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetraol

  • MDL number MFCD00061632
  • Cas Number : 54325-28-9
  • Molecular Weight : 422.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/54325-28-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/54325-28-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart