AMBH303C5E74 Ambeed, Inc.

3-(3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine

  • MDL number MFCD00242755
  • Cas Number : 303-49-1
  • Molecular Weight : 314.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 91
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 275.6�t 760 mmHg
Product Link https://www.ambeed.com/products/303-49-1.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/303-49-1.html
pictograms GHS05
hstatements H227-H314
Availability details for
Quantity
  • Availability
  • Add to Cart