AMBH303C5E7A Ambeed, Inc.

Methyl 3-amino-5-fluoropicolinate

  • MDL number MFCD18259055
  • Cas Number : 1052714-11-0
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1052714-11-0.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/1052714-11-0.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart