AMBH303C5E93 Ambeed, Inc.

(Z)-3,5-Dichloro-4-(3-ethoxy-2-methyl-3-oxoprop-1-en-1-yl)benzoic acid

  • MDL number MFCD30471709
  • Molecular Weight : 303.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/P000346659.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/P000346659.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart