AMBH303C5EA8 Ambeed, Inc.

2-Methylpropane-1,2-diamine

  • MDL number MFCD00008054
  • Cas Number : 811-93-8
  • Molecular Weight : 88.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 23.9
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 130.9�t 760 mmHg
Product Link https://www.ambeed.com/products/811-93-8.html

Safety Information

signalword Danger
pcodes P210-P233-P240-P241+P242+P243-P260-P264-P280-P301+P330+P331+P310-P303+P361+P353+P310+P363-P304+P340+P310-P305+P351+P338+P310-P370+P378-P403+P235-P405-P501
hcodes 8,3
MSDS https://www.ambeed.com/products/811-93-8.html
pictograms GHS02,GHS05
hstatements H226-H314
Availability details for
Quantity
  • Availability
  • Add to Cart