AMBH303C5EC4 Ambeed, Inc.

H-Orn(2-Cl-Z)-OH

  • MDL number MFCD00235874
  • Cas Number : 118553-99-4
  • Molecular Weight : 300.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-2-Amino-5-((((2-chlorobenzyl)oxy)carbonyl)amino)pentanoic acid
Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 516.7�t 760 mmHg
Product Link https://www.ambeed.com/products/118553-99-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/118553-99-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart