AMBH303C5ECA Ambeed, Inc.

tert-Butyl 3-oxo-4-(2-oxo-2-(4-(trifluoromethyl)phenyl)ethyl)-1,4-diazepane-1-carboxylate

  • MDL number MFCD30478725
  • Cas Number : 1998216-12-8
  • Molecular Weight : 400.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1998216-12-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1998216-12-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart