AMBH303C5EE6 Ambeed, Inc.

10-Undecenoic Acid Zinc

  • MDL number MFCD00067246
  • Cas Number : 557-08-4
  • Molecular Weight : 431.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/557-08-4.html

Safety Information

signalword Danger
pcodes P273-P305+P351+P338
MSDS https://www.ambeed.com/products/557-08-4.html
pictograms GHS07,GHS09
hstatements H302-H315-H319-H410
Availability details for
Quantity
  • Availability
  • Add to Cart