AMBH303C5EF4 Ambeed, Inc.

3-Iodopyrazine-2-carboxylic acid

  • MDL number MFCD13193281
  • Cas Number : 212471-40-4
  • Molecular Weight : 250

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/212471-40-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/212471-40-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart