AMBH303C5EFC Ambeed, Inc.

4-([2,2':6',2''-Terpyridin]-4'-yl)aniline

  • MDL number MFCD06796989
  • Cas Number : 178265-65-1
  • Molecular Weight : 324.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/178265-65-1.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/178265-65-1.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart