AMBH303C5F05 Ambeed, Inc.

2-Phenyldihydro-2H-pyran-4(3H)-one

  • MDL number MFCD11112124
  • Cas Number : 147688-62-8
  • Molecular Weight : 176.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/147688-62-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/147688-62-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart