AMBH303C5F19 Ambeed, Inc.

5-Bromo-6-methylnicotinic acid

  • MDL number MFCD18257500
  • Cas Number : 1190862-72-6
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1190862-72-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1190862-72-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart