AMBH303C5F58 Ambeed, Inc.

HJC0350

  • MDL number MFCD05144768
  • Cas Number : 885434-70-8
  • Molecular Weight : 277.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Mesitylsulfonyl)-2,4-dimethyl-1H-pyrrole
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/885434-70-8.html

Safety Information

signalword Danger
pcodes P273
hcodes 9
MSDS https://www.ambeed.com/products/885434-70-8.html
pictograms GHS07,GHS09
hstatements H302-H410
Availability details for
Quantity
  • Availability
  • Add to Cart