AMBH303C5F70 Ambeed, Inc.

Uridine 5'-(trihydrogen diphosphate) sodium salt

  • MDL number N/A
  • Cas Number : 1457-11-0
  • Molecular Weight : 426.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1457-11-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1457-11-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart