AMBH303C5F77 Ambeed, Inc.

3-(Benzyloxy)isoxazole-5-carboxylic acid

  • MDL number MFCD11499028
  • Cas Number : 2552-54-7
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-120
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 441.6�0�t 760 mmHg
Product Link https://www.ambeed.com/products/2552-54-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2552-54-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart