AMBH303C5FAC Ambeed, Inc.

Methyl 3-amino-5-chloro-2-methylbenzoate

  • MDL number MFCD23701503
  • Cas Number : 294190-18-4
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/294190-18-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/294190-18-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart