AMBH303C5FFF Ambeed, Inc.

Methyl quinoline-7-carboxylate

  • MDL number MFCD18417763
  • Cas Number : 51552-68-2
  • Molecular Weight : 187.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/51552-68-2.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/51552-68-2.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart