AMBH303C6030 Ambeed, Inc.

2-(4-Nitrophenyl)thiazole

  • MDL number MFCD26794836
  • Cas Number : 3704-41-4
  • Molecular Weight : 206.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3704-41-4.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/3704-41-4.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart