AMBH303C6051 Ambeed, Inc.

3-Bromothieno[3,2-c]pyridin-4(5H)-one

  • MDL number MFCD14706352
  • Cas Number : 799293-83-7
  • Molecular Weight : 230.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/799293-83-7.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/799293-83-7.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart