AMBH303C6094 Ambeed, Inc.

1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid

  • MDL number MFCD11041143
  • Cas Number : 756526-02-0
  • Molecular Weight : 663.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 781.1�0�t 760 mmHg
Product Link https://www.ambeed.com/products/756526-02-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/756526-02-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart