AMBH303C6099 Ambeed, Inc.

(2-(Difluoromethoxy)pyridin-4-yl)methanamine hydrochloride

  • MDL number MFCD21605147
  • Cas Number : 943843-27-4
  • Molecular Weight : 210.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/943843-27-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/943843-27-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart