AMBH303C60B3 Ambeed, Inc.

5-Bromo-2-(carboxymethyl)benzoic acid

  • MDL number MFCD11877995
  • Cas Number : 19725-82-7
  • Molecular Weight : 259.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/19725-82-7.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/19725-82-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart