AMBH303C60BA Ambeed, Inc.

3,4-Dichloro-1H-pyrrolo[3,2-c]pyridine

  • MDL number MFCD12963262
  • Cas Number : 117332-47-5
  • Molecular Weight : 187.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/117332-47-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/117332-47-5.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart