AMBH303C60E5 Ambeed, Inc.

Diphenmanil methylsulfate

  • MDL number MFCD00865157
  • Cas Number : 62-97-5
  • Molecular Weight : 389.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/62-97-5.html

Safety Information

signalword Danger
pcodes P261-P280-P301+P310-P305+P351+P338-P342+P311
MSDS https://www.ambeed.com/products/62-97-5.html
pictograms GHS05,GHS06,GHS08
hstatements H301-H318-H334
Availability details for
Quantity
  • Availability
  • Add to Cart