AMBH303C6109 Ambeed, Inc.

2,5-Dimethyl-1,3,4-oxadiazole

  • MDL number MFCD00483011
  • Cas Number : 13148-65-7
  • Molecular Weight : 98.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/13148-65-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/13148-65-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart