AMBH303C611E Ambeed, Inc.

7-Amino-4,5-dihydro-5-oxo-pyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD11040171
  • Cas Number : 138904-35-5
  • Molecular Weight : 175.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 290
Purity 97
Flash Point 156.617
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/138904-35-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/138904-35-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart