AMBH303C6136 Ambeed, Inc.

tert-Butyl 6-amino-1H-indazole-1-carboxylate

  • MDL number MFCD04114657
  • Cas Number : 219503-81-8
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 395.3�t 760 mmHg
Product Link https://www.ambeed.com/products/219503-81-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/219503-81-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart