AMBH303C613D Ambeed, Inc.

Pentaethylene Glycol Monomethyl Ether

  • MDL number MFCD00045995
  • Cas Number : 23778-52-1
  • Molecular Weight : 252.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 154.7
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 332.3�t 760 mmHg
Product Link https://www.ambeed.com/products/23778-52-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/23778-52-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart