AMBH303C614A Ambeed, Inc.

Methyl 3-(tert-butyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00829900
  • Cas Number : 1017782-45-4
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1017782-45-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1017782-45-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart