AMBH303C614B Ambeed, Inc.

2-Methyloxazole

  • MDL number MFCD08669512
  • Cas Number : 23012-10-4
  • Molecular Weight : 83.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/23012-10-4.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338
hcodes 8
MSDS https://www.ambeed.com/products/23012-10-4.html
pictograms GHS02,GHS05
hstatements H226-H318
Availability details for
Quantity
  • Availability
  • Add to Cart