AMBH303C616B Ambeed, Inc.

Ethyl 7-(furan-2-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylate

  • MDL number N/A
  • Cas Number : 102269-47-6
  • Molecular Weight : 283.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/102269-47-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/102269-47-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart