AMBH303C616F Ambeed, Inc.

4-Chloro-6-(chloromethyl)-2-methoxypyrimidine

  • MDL number MFCD28400977
  • Cas Number : 94170-69-1
  • Molecular Weight : 193.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 140.6
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 308.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/94170-69-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/94170-69-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart