AMBH303C6185 Ambeed, Inc.

2-Iodo-4-methoxybenzoic acid

  • MDL number MFCD09909619
  • Cas Number : 54435-09-5
  • Molecular Weight : 278.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 361.0�0�t 760 mmHg
Product Link https://www.ambeed.com/products/54435-09-5.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/54435-09-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart