AMBH303C6195 Ambeed, Inc.

3-Chloro-2-methylimidazo[1,2-b]pyridazine

  • MDL number N/A
  • Cas Number : 1935233-51-4
  • Molecular Weight : 167.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1935233-51-4.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1935233-51-4.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart