AMBH303C61B0 Ambeed, Inc.

4''-Pentyl-[1,1':4',1''-terphenyl]-4-carbonitrile

  • MDL number MFCD00799423
  • Cas Number : 54211-46-0
  • Molecular Weight : 325.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/54211-46-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/54211-46-0.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart