AMBH303C61E9 Ambeed, Inc.

9-(Naphthalen-1-yl)-9H-carbazole

  • MDL number MFCD26127424
  • Cas Number : 22034-43-1
  • Molecular Weight : 293.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/22034-43-1.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/22034-43-1.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart