AMBH303C61F9 Ambeed, Inc.

2-Methoxy-4-(trifluoromethyl)pyridine

  • MDL number MFCD00096939
  • Cas Number : 219715-34-1
  • Molecular Weight : 177.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/219715-34-1.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/219715-34-1.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart